missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Aminosulfonyl)-4-chlorobenzoyl]-benzonic Acid, TRC

Product Code. 30179932
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179932 250 mg 250mg
30180319 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179932 Supplier Toronto Research Chemicals Supplier No. TRCA633000250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5270-74-6
Molecular Formula C14 H10 Cl N O5 S
Molecular Weight (g/mol) 339.75
Synonym 2-[3-(Aminosulfonyl)-4-chlorobenzoyl]benzoic acid, o-(4-Chloro-3-sulfamoylbenzoyl)benzoic acid
IUPAC Name 2-(4-chloro-3-sulfamoylbenzoyl)benzoic acid
SMILES NS(=O)(=O)c1cc(ccc1Cl)C(=O)c2ccccc2C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H10ClNO5S/c15-11-6-5-8(7-12(11)22(16,20)21)13(17)9-3-1-2-4-10(9)14(18)19/h1-7H,(H,18,19)(H2,16,20,21)
Formula Weight 338.9968
Percent Purity >95%
Chemical Name or Material 2-(4-Chloro-3-Sulfamoylbenzoyl)Benzoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.