missing translation for 'onlineSavingsMsg'
Learn More

2,3,7,8-Tetrachloro-p-dioxin, TRC

Product Code. 30422336
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 30422336

Brand: Toronto Research Chemicals TRCT2913701MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,7,8-Tetrachlorodibenzo-p-dioxin
CAS 1746-01-6
Purity Grade Notes HPLC
Molecular Formula C12 H4 Cl4 O2
Quantity 1 mg
Synonym Dibenzo[b,e][1,4]dioxin','2,3,7,8-tetrachloro-, Dibenzo-p-dioxin','2,3,7,8-tetrachloro- (6CI,7CI,8CI), 2,3,7,8-Tetrachlorodibenzo[b,e][1,4]dioxin, 2,3,7,8-TCDD, 2,3,7,8-Tetrachloro-p-dioxin, 2,3,7,8-Tetrachlorodibenzo-1,4-dioxin, 2,3,7,8-Tetrachlorodibenzo-p-dioxin, 2,3,7,8-Tetrachlorodibenzodioxin, 2,3,7,8-tetrachlorodibenzo-p-dioxin, 2378TCDD, D 48, Dioxin, Dioxin (herbicide contaminant), PCDD 48, TCDBD, TCDD, Tetrachlorodibenzo-p-dioxin, Tetrachlorodibenzodioxin
InChI Formula InChI=1S/C12H4Cl4O2/c13-5-1-9-10(2-6(5)14)18-12-4-8(16)7(15)3-11(12)17-9/h1-4H
SMILES Clc1cc2Oc3cc(Cl)c(Cl)cc3Oc2cc1Cl
IUPAC Name 2,3,7,8-tetrachlorodibenzo-p-dioxin
Molecular Weight (g/mol) 321.97
Formula Weight 319.8965
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.