missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,6-Trifluorotoluene, 98%, Thermo Scientific™
£10.80 - £151.00
Chemical Identifiers
CAS | 119916-25-5 |
---|---|
Molecular Formula | C7H5Cl3 |
Molecular Weight (g/mol) | 195.467 |
MDL Number | MFCD12922564 |
InChI Key | UZYYBZNZSSNYSA-UHFFFAOYSA-N |
Synonym | 2,3,6-trichlorotoluene, benzene, 1,2,4-trichloro-3-methyl, toluene, 2,3,6-trichloro, unii-19m2l15z6u, 2,3,6-trichorotoluene, 2,6-trichlorotoluene, toluene,3,6-trichloro, 2,3,6-trichloro-toluen, toluene,2,3,6-trichloro, 4-05-00-00819 beilstein handbook reference |
PubChem CID | 16378 |
IUPAC Name | 1,2,4-trichloro-3-methylbenzene |
SMILES | CC1=C(C=CC(=C1Cl)Cl)Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
119916-25-5 | |
195.467 | |
UZYYBZNZSSNYSA-UHFFFAOYSA-N | |
16378 | |
CC1=C(C=CC(=C1Cl)Cl)Cl |
C7H5Cl3 | |
MFCD12922564 | |
2,3,6-trichlorotoluene, benzene, 1,2,4-trichloro-3-methyl, toluene, 2,3,6-trichloro, unii-19m2l15z6u, 2,3,6-trichorotoluene, 2,6-trichlorotoluene, toluene,3,6-trichloro, 2,3,6-trichloro-toluen, toluene,2,3,6-trichloro, 4-05-00-00819 beilstein handbook reference | |
1,2,4-trichloro-3-methylbenzene |
Specifications
119916-25-5 | |
Liquid | |
MFCD12922564 | |
UZYYBZNZSSNYSA-UHFFFAOYSA-N | |
1,2,4-trichloro-3-methylbenzene | |
16378 | |
98% |
Colorless to Yellow | |
C7H5Cl3 | |
2,3,6-trichlorotoluene, benzene, 1,2,4-trichloro-3-methyl, toluene, 2,3,6-trichloro, unii-19m2l15z6u, 2,3,6-trichorotoluene, 2,6-trichlorotoluene, toluene,3,6-trichloro, 2,3,6-trichloro-toluen, toluene,2,3,6-trichloro, 4-05-00-00819 beilstein handbook reference | |
CC1=C(C=CC(=C1Cl)Cl)Cl | |
195.467 | |
146.11 | |
2,3,6-Trifluorotoluene |