missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,6,7,12,13,16,17-Octahydro-9-[4-[[[2-[(methylsulfonyl)thio]ethyl]amino]sulfonyl]-2-sulfophenyl]-1H,5H,11H,15H-xantheno[2,3,4-ij:5,6,7-i'j′]diquinolizin-18-ium inner salt, TRC
Specifications
Specifications
| Chemical Name or Material | Texas Red-2-sulfonamidoethyl methanethiosulfonate |
| CAS | 386229-76-1 |
| Molecular Formula | C34 H37 N3 O8 S4 |
| Quantity | 25 mg |
| Synonym | 1H,5H,11H,15H-Xantheno[2,3,4-ij:5,6,7-i'j']diquinolizin-18-ium','2,3,6,7,12,13,16,17-octahydro-9-[4-[[[2-[(methylsulfonyl)thio]ethyl]amino]sulfonyl]-2-sulfophenyl]-','inner salt (9CI), Texas Red-2-sulfonamidoethyl methanethiosulfonate |
| InChI Formula | InChI=1S/C34H37N3O8S4/c1-47(38,39)46-17-12-35-48(40,41)23-10-11-24(29(20-23)49(42,43)44)30-27-18-21-6-2-13-36-15-4-8-25(31(21)36)33(27)45-34-26-9-5-16-37-14-3-7-22(32(26)37)19-28(30)34/h10-11,18-20,35H,2-9,12-17H2,1H3 |
| SMILES | CS(=O)(=O)SCCNS(=O)(=O)c1ccc(c2c3cc4CCCN5CCCc(c45)c3[o+]c6c7CCCN8CCCc(cc26)c78)c(c1)S(=O)(=O)[O-] |
| Molecular Weight (g/mol) | 743.93 |
| Formula Weight | 743.1463 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?