missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triphenyl-2H-tetrazolium chloride, 98%

Product Code. 11474213
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474213 50 g 50g
11464213 10 g 10g
11484213 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474213 Supplier Thermo Scientific Alfa Aesar Supplier No. A10870.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, C19H15ClN4, CAS Number-298-96-4, triphenyltetrazolium chloride, tetrazolium chloride, urocheck, tetrazolium red, tetrazolium chloride, uroscreen, red tetrazolium, vitastain, 2,3,5-triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride | CAS: 298-96-4 | C19H15ClN4 | 334.81 g/mol

2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.

Solubility
Soluble in water, ethanol and acetone. Insoluble in ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-96-4
Molecular Formula C19H15ClN4
Molecular Weight (g/mol) 334.81
MDL Number MFCD00011963
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
Synonym 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride
PubChem CID 9283
ChEBI CHEBI:78019
IUPAC Name 2,3,5-triphenyltetrazol-2-ium;chloride
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽250°C (decomposition)
Assay Percent Range 98%
Quantity 50 g
Beilstein 3923356
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9744
Solubility Information Soluble in water,ethanol and acetone. Insoluble in ether.
Formula Weight 334.81
Percent Purity 98%
Chemical Name or Material 2, 3, 5-Triphenyl-2H-tetrazolium chloride
compliance-icons
Product Identifier
  • 2,3,5-Triphenyl-2H-tetrazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,3,5-triphenyl-2H-Tetrazolium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.