missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylphenol, 98+%

Product Code. 11409632
Change view
Click to view available options
Quantity:
25 g
100 g
1000 g
Unit Size:
1000g
100g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409632 1000 g 1000g
11494110 25 g 25g
11404120 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409632 Supplier Thermo Scientific Alfa Aesar Supplier No. A10745.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 697-82-5 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-82-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002228
InChI Key OGRAOKJKVGDSFR-UHFFFAOYSA-N
Synonym isopseudocumenol, phenol, 2,3,5-trimethyl, 1-hydroxy-2,3,5-trimethylbenzene, unii-1s1061zbq1, phenol, 2,3,5 or 3,4,5-trimethyl, pubchem2249, 2,3,5-trimethyphenol, phenol,3,5-trimethyl, 2,3,5-trimethyl-phenol, 6-hydroxypseudocumene
PubChem CID 12769
ChEBI CHEBI:38570
IUPAC Name 2,3,5-trimethylphenol
SMILES CC1=CC(=C(C(=C1)O)C)C

Specifications

Melting Point 92°C to 95°C
Boiling Point 230°C to 231°C
Odor Phenol-like
Quantity 1000 g
UN Number UN2430
Beilstein 2042210
Formula Weight 136.2
Percent Purity ≥98%
Chemical Name or Material 2,3,5-Trimethylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.