missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylfuran, 99%

Product Code. 15214358 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15214358 5 g 5g
15204358 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15214358 Supplier Thermo Scientific Acros Supplier No. 454470050
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10504-04-8 | C7H10O | 110.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10504-04-8
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD03931179
InChI Key NJXZFRUNHWKHEC-UHFFFAOYSA-N
Synonym furan, 2,3,5-trimethyl, 2,3,5-trimethyl-furan, furan,2,3,5-trimethyl
PubChem CID 517859
IUPAC Name 2,3,5-trimethylfuran
SMILES CC1=CC(C)=C(C)O1

Specifications

Density 0.8900g/mL
Boiling Point 121.0°C to 122.0°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Refractive Index 1.449
Quantity 5 g
Specific Gravity 0.89
Formula Weight 110.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2,3,5-Trimethylfuran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3,5-Trimethylfuran, 99%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.