missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triiodobenzoic Acid, TRC

Product Code. 30138713
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138713 10 g 10g
30006968 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138713 Supplier Toronto Research Chemicals Supplier No. TRCT79541510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-82-4
Molecular Formula C7 H3 I3 O2
Molecular Weight (g/mol) 499.81
Synonym 2,3,5-Triiodobenzoic Acid, Benzoic acid, 2,3,5-triiodo- (8CI, 9CI, ACI), 2,3,5-Triiodobenzoic acid (ACI), 2,3,5-TIBA, A 20812, Floraltone, Johnkolor, NSC 2582, TIB, TIBA, Triiodobenzoic acid
IUPAC Name 2,3,5-triiodobenzoic acid
SMILES OC(=O)c1cc(I)cc(I)c1I

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H3I3O2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,(H,11,12)
Formula Weight 499.7267
Chemical Name or Material 2,3,5-Triiodobenzoic acid
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.