missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triiodobenzoic acid, 98+%

Product Code. 11397785
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397785 25 g 25g
11387785 5 g 5g
11307795 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397785 Supplier Thermo Scientific Alfa Aesar Supplier No. L02679.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-82-4 | C7H3I3O2 | 499.81 g/mol

2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-82-4
Molecular Formula C7H3I3O2
Molecular Weight (g/mol) 499.81
MDL Number MFCD00002420
InChI Key ZMZGFLUUZLELNE-UHFFFAOYSA-N
Synonym floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova
PubChem CID 6948
ChEBI CHEBI:73175
IUPAC Name 2,3,5-triiodobenzoic acid
SMILES OC(=O)C1=CC(I)=CC(I)=C1I

Specifications

Melting Point 220°C to 222°C
Quantity 25 g
Beilstein 1955088
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 499.81
Percent Purity ≥98%
Chemical Name or Material 2,3,5-Triiodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.