missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trifluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number 11491300
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
654-87-5
C7H3F3O2
176.09
MFCD00061200
CPZROMDDCPPFOO-UHFFFAOYSA-N
2,3,5-trilfluorobenzoic acid, benzoic acid, 2,3,5-trifluoro, pubchem1401, acmc-209nr1, 2,3,5-trifluorobenzoicacid, ksc622o6j, 2,3,5-trifluoro-benzoic acid, rarechem al bo 0710, benzoic acid,2,3,5-trifluoro, cpzromddcppfoo-uhfffaoysa
2776991
2,3,5-trifluorobenzoic acid
OC(=O)C1=CC(F)=CC(F)=C1F
This item is not returnable. View return policy
654-87-5
C7H3F3O2
176.09
MFCD00061200
CPZROMDDCPPFOO-UHFFFAOYSA-N
2,3,5-trilfluorobenzoic acid, benzoic acid, 2,3,5-trifluoro, pubchem1401, acmc-209nr1, 2,3,5-trifluorobenzoicacid, ksc622o6j, 2,3,5-trifluoro-benzoic acid, rarechem al bo 0710, benzoic acid,2,3,5-trifluoro, cpzromddcppfoo-uhfffaoysa
2776991
2,3,5-trifluorobenzoic acid
OC(=O)C1=CC(F)=CC(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 108°C to 114°C
Quantity 5 g
Formula Weight 176.1
Percent Purity 97%
Chemical Name or Material 2,3,5-Trifluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'