missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trifluoroaniline, 97%, Thermo Scientific™

Catalog Number 15493547
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
363-80-4
C6H4F3N
147.1
MFCD00083549
AMGDNQWQBWPRPR-UHFFFAOYSA-N
2776933
2,3,5-trifluoroaniline
C1=C(C=C(C(=C1F)F)N)F
This item is not returnable. View return policy
363-80-4
C6H4F3N
147.1
MFCD00083549
AMGDNQWQBWPRPR-UHFFFAOYSA-N
2776933
2,3,5-trifluoroaniline
C1=C(C=C(C(=C1F)F)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.4895
Quantity 250 mg
UN Number UN2941
Synonym benzenamine, 2,3,5-trifluoro, 2,3,5-trifluoro aniline, 2,3,5-trifluoro-phenylamine, pubchem1549, acmc-1cnbv, intermediates-zcf02613, 2,3,5-trifluorophenylamine, ksc576g7j, benzenamine,2,3,5-trifluoro
Formula Weight 147.1
Percent Purity 97%
Chemical Name or Material 2,3,5-Trifluoroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'