missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichlorotoluene, TRC

Product Code. 17901055
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901055 250 mg 250mg
17944485 1 g 1g
17964785 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17901055 Supplier Toronto Research Chemicals Supplier No. TRCT353260250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56961-86-5
Molecular Formula C7 H5 Cl3
Molecular Weight (g/mol) 195.474
Synonym 1,2,5-Trichloro-3-methylbenzene (ACI), 2,3,5-Trichlorotoluene, 1,2,5-Trichloro-3-methylbenzene
IUPAC Name 1,2,5-trichloro-3-methylbenzene
SMILES Cc1cc(Cl)cc(Cl)c1Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H5Cl3/c1-4-2-5(8)3-6(9)7(4)10/h2-3H,1H3
Formula Weight 193.9457
Percent Purity >95%
Chemical Name or Material 2,3,5-Trichlorotoluene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.