missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichloropyridine, Thermo Scientific™

Product Code. 10693025 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 10693025

Brand: Thermo Scientific Acros 247020500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16063-70-0
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.44
MDL Number MFCD00043007
InChI Key CNLIIAKAAMFCJG-UHFFFAOYSA-N
Synonym pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
PubChem CID 27666
IUPAC Name 2,3,5-trichloropyridine
SMILES C1=C(C=NC(=C1Cl)Cl)Cl

Specifications

Melting Point 46°C to 50°C
Boiling Point 219°C
Flash Point 113°C
Packaging Glass bottle
Quantity 50 g
Formula Weight 182.44
Chemical Name or Material 2, 3, 5-Trichloropyridine
Product Identifier
  • 2,3,5-Trichloropyridine
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.