missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone, TRC

Product Code. 30450357
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30450357 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30450357

Brand: Toronto Research Chemicals TRCT7713785G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone
CAS 55094-52-5
Molecular Formula C26 H26 O5
Quantity 5 g
Synonym Remdesivir Impurity 21
InChI Formula InChI=1S/C26H26O5/c27-26-25(30-18-22-14-8-3-9-15-22)24(29-17-21-12-6-2-7-13-21)23(31-26)19-28-16-20-10-4-1-5-11-20/h1-15,23-25H,16-19H2/t23-,24-,25-/m1/s1
SMILES O=C1O[C@H](COCc2ccccc2)[C@@H](OCc3ccccc3)[C@H]1OCc4ccccc4
IUPAC Name (3R,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-one
Molecular Weight (g/mol) 418.482
Formula Weight 418.178
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.