missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-beta-D-arabinofuranose

Product Code. 15488379
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488379 10 g 10g
15468379 1 g 1g
15478379 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488379 Supplier Thermo Scientific Alfa Aesar Supplier No. J66853.09
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60933-68-8 | C26H28O5 | 420.505 g/mol

An arabinofuranose derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An arabinofuranose derivative.

Solubility
Soluble in methanol

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60933-68-8
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD00047076
InChI Key NAQUAXSCBJPECG-FXSWLTOZSA-N
Synonym 2,3,5-tri-o-benzyl-beta-d-arabinofuranose, 2,3,5-tri-o-benzyl-b-d-arabinofuranose, 2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2-o,3-o,5-o-tribenzyl-beta-d-arabinofuranose, 2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
PubChem CID 11729473
IUPAC Name (2R,3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Color White
Quantity 10 g
Solubility Information Soluble in methanol
Formula Weight 420.5
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-beta-D-arabinofuranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.