missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
10 g
missing translation for 'unitSize'
10g
1g
5g
Descrizione
An arabinofuranose derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
An arabinofuranose derivative.
Solubility
Soluble in methanol
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
An arabinofuranose derivative.
Solubility
Soluble in methanol
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifica
Specifica
| Color | White |
| Quantity | 5 g |
| Solubility Information | Soluble in methanol |
| Formula Weight | 420.5 |
| Physical Form | Powder |
| Chemical Name or Material | 2,3,5-Tri-O-benzyl-beta-D-arabinofuranose |
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto