missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-1-O-(4-nitrobenzoyl)-D-arabinofuranose, 98%, Thermo Scientific Chemicals

Product Code. 15430517
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430517 5 g 5g
15420517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430517 Supplier Thermo Scientific Alfa Aesar Supplier No. J66165.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52522-49-3
Molecular Formula C33H31NO8
Molecular Weight (g/mol) 569.61
MDL Number MFCD00038477
InChI Key VBDBDZHLJKDSSB-UFRUDBOASA-N
Synonym 2,3,5-tri-o-benzyl-1-o-4-nitrobenzoyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-1-o-p-nitrobenzoyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-d-arabinofuranose 1-4-nitrobenzoate, d-arabinofuranose,2,3,5-tris-o-phenylmethyl-, 4-nitrobenzoate 9ci, 3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-yl 4-nitrobenzoate, 3s,4s,5r-3,4-bis phenylmethoxy-5-phenylmethoxy methyl oxolan-2-yl 4-nitro benzoate, 1-o-4-nitrobenzoyl-2,3,5-tri-o-benzyl-d-arabinofuranose, 4-nitrobenzoate 2,3,5-tris-o-benzyl-d-arabinofuranose, 3s,4r,5r-3,4-dibenzyloxy-5-benzyloxymethyl tetrahydrofuran-2-yl 4-nitrobenzoate
PubChem CID 97076
IUPAC Name [(3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-yl] 4-nitrobenzoate
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)OC(=O)C3=CC=C(C=C3)[N+](=O)[O-])OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Melting Point 69°C to 72°C
Color White
Quantity 5 g
Beilstein 1303859
Formula Weight 569.6
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-1-O-(4-nitrobenzoyl)-D-arabinofuranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.