missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) iodide, ultra dry, 99.999% (metals basis)

Product Code. 11388527
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388527 5 g 5g
11398527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388527 Supplier Thermo Scientific Alfa Aesar Supplier No. 035699.06
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 13450-91-4 | GaI3 | 450.436 g/mol

Gallium(III) iodide is widely used as lewis acid, a precursor of organogallium reagents. It can also be involved in detection of solar neutrinos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-91-4
Molecular Formula GaI3
Molecular Weight (g/mol) 450.436
MDL Number MFCD00016107
InChI Key DWRNSCDYNYYYHT-UHFFFAOYSA-K
Synonym gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83478
IUPAC Name triiodogallane
SMILES [Ga](I)(I)I

Specifications

Boiling Point 345°C (sublimation)
Melting Point 212°C
Color Yellow to Green
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in aromatic solvents.
Formula Weight 450.43
Sensitivity Light and moisture sensitive
Appearance Yellow-green powder
Density 4.15
Chemical Name or Material Gallium(III) iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.