missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-difluorophenyl)acetic acid, Maybridge™

Brand:  Maybridge SB01687FA

Code : 31

Additional Details : CAS Number : 105184-38-1 Weight : 0.15000kg

Quantity 100g

 View more versions of this product

Product Code. 10082364

  • £120.00 / 100g

Estimated Shipment
13-01-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

105184-38-1
172.131
3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
2-(3,5-difluorophenyl)acetic acid
C8H6F2O2
IGGNSAVLXJKCNH-UHFFFAOYSA-N
145424
C1=C(C=C(C=C1F)F)CC(=O)O

Specifications

2-(3, 5-Difluorophenyl)acetic acid
100g
97%
3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
C1=C(C=C(C=C1F)F)CC(=O)O
172.131
172.13
Amber glass bottle
105184-38-1
C8H6F2O2
IGGNSAVLXJKCNH-UHFFFAOYSA-N
2-(3,5-difluorophenyl)acetic acid
145424
97%
Videos
SDS Documents