missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Collidine, 99+%

Product Code. 10033290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10033290 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10033290 Supplier Thermo Scientific Acros Supplier No. 275470250
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 695-98-7 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 695-98-7
CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 182°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C8H11N
Refractive Index 1.5065 to 1.5085
MDL Number MFCD00126840
Quantity 25 g
Specific Gravity 0.93
Synonym 2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ether,acetone
InChI Key GFYHSKONPJXCDE-UHFFFAOYSA-N
SMILES CC1=CC(=C(N=C1)C)C
IUPAC Name 2,3,5-trimethylpyridine
Molecular Weight (g/mol) 121.18
PubChem CID 12759
Formula Weight 121.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2, 3, 5-Collidine, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3,5-Collidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.