missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Collidine, 99%, Thermo Scientific Chemicals

Catalog Number 11455547
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
695-98-7
C8H11N
121.183
MFCD00126840
GFYHSKONPJXCDE-UHFFFAOYSA-N
2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f
12759
2,3,5-trimethylpyridine
CC1=CC(=C(N=C1)C)C
This item is not returnable. View return policy
695-98-7
C8H11N
121.183
MFCD00126840
GFYHSKONPJXCDE-UHFFFAOYSA-N
2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f
12759
2,3,5-trimethylpyridine
CC1=CC(=C(N=C1)C)C

2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Density 0.935
Boiling Point 182°C to 184°C
Flash Point 74°C (165°F)
Odor Unpleasant
Refractive Index 1.5075
Quantity 5 g
UN Number UN2810
Beilstein 108832
Solubility Information Slightly soluble in water.
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,3,5-Collidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'