missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethylpyrazine, 98+%

Product Code. 11463827
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463827 100 g 100g
11453827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463827 Supplier Thermo Scientific Alfa Aesar Supplier No. A18279.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1124-11-4 | C8H12N2 | 136.198 g/mol

2,3,5,6-Tetramethylpyrazine is a flavor ingredient of tobacco. It forms cocrystal with 4-hydroxybenzoic acid which exhibits supramolecular synthon polymorphism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5,6-Tetramethylpyrazine is a flavor ingredient of tobacco. It forms cocrystal with 4-hydroxybenzoic acid which exhibits supramolecular synthon polymorphism.

Solubility
Soluble in water (4 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-11-4
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00006146
InChI Key FINHMKGKINIASC-UHFFFAOYSA-N
Synonym tetramethylpyrazine, ligustrazine, pyrazine, tetramethyl, bs factor, tetramethylpyrazin, liqustrazine, tetrapyrazine, 2,3,5,6-tetramethyl pyrazine, chuanxiongzine, ligustizine
PubChem CID 14296
IUPAC Name 2,3,5,6-tetramethylpyrazine
SMILES CC1=C(N=C(C(=N1)C)C)C

Specifications

Melting Point 83°C to 86°C
Density 1.08
pH 7
Boiling Point 190°C
Odor Pleasant
Quantity 100 g
Beilstein 113100
Solubility Information Soluble in water (4g/L at 20°C).
Formula Weight 136.2
Percent Purity ≥98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetramethylpyrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.