missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethylbenzyl chloride, 98+%, Thermo Scientific™

Product Code. 11310074
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310074 5 g 5g
11320074 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310074 Supplier Thermo Scientific Alfa Aesar Supplier No. L00501.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7435-83-8
Molecular Formula C11H15Cl
Molecular Weight (g/mol) 182.691
MDL Number MFCD00013679
InChI Key UGAPPXGBBWAIGT-UHFFFAOYSA-N
Synonym 3-chloromethyl-1,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethylbenzyl chloride, benzene, 3-chloromethyl-1,2,4,5-tetramethyl, 4-05-00-01111 beilstein handbook reference, 2,3,5,6-tetramethylbenzylchloride, 2,3,5,6-tetramethyl benzyl chloride, 2,3,5,6-tetramethyl-benzyl chloride, 3-chloromethyl-1,2,4,5-tetramethyl-benzene, benzene,3-chloromethyl-1,2,4,5-tetramethyl
PubChem CID 81939
IUPAC Name 3-(chloromethyl)-1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C(=C1C)CCl)C)C

Specifications

Melting Point 68°C to 71°C
Quantity 5 g
UN Number UN3261
Beilstein 1863560
Formula Weight 182.69
Percent Purity ≥98%
Chemical Name or Material 2,3,5,6-Tetramethylbenzyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.