missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethylbenzenesulfonyl chloride, 98%

Product Code. 11490903
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490903 50 g 50g
11480903 10 g 10g
11400913 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490903 Supplier Thermo Scientific Alfa Aesar Supplier No. A13313.18
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60706-63-0 | C10H13ClO2S | 232.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60706-63-0
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.72
MDL Number MFCD00014723
InChI Key ZCXRROBIIMQMHR-UHFFFAOYSA-N
Synonym 2,3,5,6-tetramethylbenzene-1-sulfonyl chloride, 2,3,5,6-tetramethylbenzenesulphonyl chloride, 2,3,5,6-tetramethyl-benzenesulfonyl chloride, chloro 2,3,5,6-tetramethylphenyl sulfone, durylsulphonyl chloride, acmc-1b645, 2,3,5,6-tetramethylbenzenesulfonylchloride, 2,3,5,6-tetramethylphenylsulfonyl chloride, benzenesulfonylchloride, 2,3,5,6-tetramethyl, benzenesulfonyl chloride, 2,3,5,6-tetramethyl-7ci, 9ci
PubChem CID 143740
IUPAC Name 2,3,5,6-tetramethylbenzenesulfonyl chloride
SMILES CC1=CC(C)=C(C)C(=C1C)S(Cl)(=O)=O

Specifications

Melting Point 97°C to 101°C
Quantity 50 g
UN Number UN3261
Beilstein 1876644
Sensitivity Moisture sensitive
Formula Weight 232.73
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetramethylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.