missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorophenylhydrazine, 97%

Product Code. 11415187
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11415187

Brand: Thermo Scientific Alfa Aesar B22600.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 653-11-2 | C6H4F4N2 | 180.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-11-2
Molecular Formula C6H4F4N2
Molecular Weight (g/mol) 180.11
MDL Number MFCD00007575
InChI Key TYMFVEOXNZYYTF-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorophenyl hydrazine, 2,3,5,6-tetrafluorphenyl hydrazin, pubchem3300, acmc-20aoi4, tymfveoxnzyytf-uhfffaoysa, 2,3,5,6-tetrafluoro-phenylhydrazine, 1-2,3,5,6-tetrafluorophenyl hydrazine, 1-hydrazino-2,3,5,6-tetrafluorobenzene, hydrazine, 2,3,5,6-tetrafluorophenyl, 1-2,3,5,6-tetrafluorophenyl hydrazine #
PubChem CID 69552
IUPAC Name (2,3,5,6-tetrafluorophenyl)hydrazine
SMILES NNC1=C(F)C(F)=CC(F)=C1F

Specifications

Melting Point 86°C to 92°C
Quantity 25 g
Beilstein 745777
Sensitivity Light sensitive
Formula Weight 180.11
Percent Purity 97%
Chemical Name or Material 2,3,5,6-Tetrafluorophenylhydrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.