missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorobenzyl bromide, 98+%

Product Code. 11381427
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381427 1 g 1g
11391427 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381427 Supplier Thermo Scientific Alfa Aesar Supplier No. L01916.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 53001-73-3 | C7H3BrF4 | 242.999 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53001-73-3
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD00013549
InChI Key QHZBSXQCGIQSID-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorobenzyl bromide, 3-bromomethyl-1,2,4,5-tetrafluorobenzene, 2,3,5,6-tetrafluorobenzylbromide, benzene, 3-bromomethyl-1,2,4,5-tetrafluoro, alpha-bromo-2,3,5,6-tetrafluorotoluene, pubchem7361, acmc-1b1z8, benzene,3-bromomethyl-1,2,4,5-tetrafluoro
PubChem CID 104377
IUPAC Name 3-(bromomethyl)-1,2,4,5-tetrafluorobenzene
SMILES C1=C(C(=C(C(=C1F)F)CBr)F)F

Specifications

Boiling Point 79°C to 80°C (80 mmHg)
Refractive Index 1.4885
Quantity 1 g
UN Number UN3265
Beilstein 2263041
Formula Weight 243
Percent Purity ≥98%
Chemical Name or Material 2,3,5,6-Tetrafluorobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.