missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorobenzeneboronic acid, 99%

Product Code. 15449109
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15449109 Supplier Thermo Scientific Alfa Aesar Supplier No. L19826.03
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 511295-01-5 | C6H3BF4O2 | 193.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 511295-01-5
Melting Point ∼140°C to 145°C
Molecular Formula C6H3BF4O2
MDL Number MFCD04039310
Quantity 1 g
Beilstein 9200167
Synonym 2,3,5,6-tetrafluorophenyl boronic acid, 2,3,5,6-tetrafluorobenzeneboronic acid, boronic acid, 2,3,5,6-tetrafluorophenyl, boronic acid, b-2,3,5,6-tetrafluorophenyl, acmc-209ks0, 2,3,5,6-tetrafluorophenylboranic acid, 2,3,5,6-tetrakis fluoranyl phenyl boronic acid, boronic acid, 2,3,5,6-tetrafluorophenyl-9ci
InChI Key SORPNEUIIZJCBF-UHFFFAOYSA-N
SMILES OB(O)C1=C(F)C(F)=CC(F)=C1F
IUPAC Name (2,3,5,6-tetrafluorophenyl)boronic acid
Molecular Weight (g/mol) 193.89
PubChem CID 2783275
Formula Weight 193.9
Percent Purity 99%
Chemical Name or Material 2,3,5,6-Tetrafluorobenzeneboronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.