missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%

Product Code. 11415757
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415757 5 g 5g
11405757 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415757 Supplier Thermo Scientific Alfa Aesar Supplier No. B24326.06
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 651-83-2 | C7H2F7N | 233.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 651-83-2
Molecular Formula C7H2F7N
Molecular Weight (g/mol) 233.089
MDL Number MFCD00091518
InChI Key FJOACTZFMHZHSC-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-4-trifluoromethyl aniline, 2,3,5,6-tetrafluoro-4-aminobenzotrifluoride, 4-amino-heptafluorotoluene, 4-amino-2,3,5,6-tetrafluorobenzotrifluoride, 4-aminotetrafluorobenzotrifluoride, 4-aminoheptafluorotoluene, 2,3,5,6-tetrafluoro-4-trifluoromethyl phenylamine, benzenamine, 2,3,5,6-tetrafluoro-4-trifluoromethyl, pubchem2817, yu-wasa auxiliary
PubChem CID 616257
IUPAC Name 2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C(F)(F)F

Specifications

Boiling Point 185°C to 187°C
Refractive Index 1.43
Quantity 5 g
UN Number UN2810
Formula Weight 233.09
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.