missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5,6-Tetrafluoro-4-methylbenzoic acid, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,3,5,6-Tetrafluoro-4-methylbenzoic Acid |
| CAS | 652-32-4 |
| Analyte or Component Names | 2,3,5,6-Tetrafluoro-4-methylbenzoic Acid |
| Shipping Condition | +20°C |
| Molecular Formula | C8 H4 F4 O2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 2,3,5,6-Tetrafluoro-4-methylbenzoic Acid, Benzoic acid, 2,3,5,6-tetrafluoro-4-methyl-, p-Toluic acid, 2,3,5,6-tetrafluoro- (7CI,8CI), 2,3,5,6-Tetrafluoro-4-methylbenzoic acid, 2,3,5,6-Tetrafluoro-p-toluic acid, 4-Methyl-2,3,5,6-tetrafluorobenzoic acid, NSC 168718 |
| InChI Formula | InChI=1S/C8H4F4O2/c1-2-4(9)6(11)3(8(13)14)7(12)5(2)10/h1H3,(H,13,14) |
| SMILES | Cc1c(F)c(F)c(C(=O)O)c(F)c1F |
| IUPAC Name | 2,3,5,6-tetrafluoro-4-methylbenzoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?