missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzoic acid, 98+%

Product Code. p-7021797
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11412437

Brand: Thermo Scientific Alfa Aesar A14278.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 573-11-5 | C10H12O5 | 212.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573-11-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00002433
InChI Key HZNQSWJZTWOTKM-UHFFFAOYSA-N
Synonym benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid
PubChem CID 11308
IUPAC Name 2,3,4-trimethoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)C(=O)O)OC)OC

Specifications

Melting Point 99°C to 102°C
Odor Odorless
Quantity 10 g
Beilstein 2696073
Formula Weight 212.2
Percent Purity ≥98%
Chemical Name or Material 2,3,4-Trimethoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.