missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzaldehyde, Mikromol™

Product Code. 30458023
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458023 4 x 25 mg 100mg
17984016 100 mg 100mg
17992776 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30458023 Supplier Mikromol Supplier No. MM0539.0300254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,3,4-Trimethoxybenzaldehyde, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,4-Trimethoxybenzaldehyde
CAS 2103-57-3
Analyte or Component Names 2,3,4-Trimethoxybenzaldehyde
Shipping Condition Room Temperature
Molecular Formula C10 H12 O4
Certifications/Compliance ISO 17025
Synonym Trimetazidine Dihydrochloride Imp. C (EP), Trimetazidine Imp. C (EP), 2,3,4-Trimethoxybenzaldehyde, Trimetazidine Dihydrochloride Impurity C, Trimetazidine Impurity C
InChI Formula InChI=1S/C10H12O4/c1-12-8-5-4-7(6-11)9(13-2)10(8)14-3/h4-6H,1-3H3
Solution Type Neat
SMILES COc1ccc(C=O)c(OC)c1OC
IUPAC Name 2,3,4-trimethoxybenzaldehyde
Molecular Weight (g/mol) 196.2
Quantity 4 x 25 mg
Formula Weight 196.0736 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.