missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzaldehyde, 99%

Product Code. 10295590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295590 50 g Glass bottle 50g
10792391 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295590 Supplier Thermo Scientific Acros Supplier No. 166450500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2103-57-3 | C10H12O4 | 196.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2103-57-3
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.2
MDL Number MFCD00003310
InChI Key UCTUXUGXIFRVGX-UHFFFAOYSA-N
Synonym trimethoxybenzaldehyde, benzaldehyde, 2,3,4-trimethoxy, 2,3,4-trimethoxy-benzaldehyde, benzaldehyde, trimethoxy, 2,3,4 trimethoxybenzylaldehyde, pubchem8261, trimethoxy benzaldehyde, trimetazidine impurity c, acmc-1b07v, ksc201q5r
PubChem CID 75006
IUPAC Name 2,3,4-trimethoxybenzaldehyde
SMILES COC1=C(C(=C(C=C1)C=O)OC)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 38.0°C to 41.0°C
Color White to Yellow
Boiling Point 168.0°C to 170.0°C (12.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3O)3C6H2CHO
Refractive Index 1.5547
Quantity 50 g
Beilstein 08, I, 684
Formula Weight 196.2
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 3, 4-Trimethoxybenzaldehyde, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.