missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzaldehyde, 98+%

Product Code. 11413304
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413304 50 g 50g
11403304 10 g 10g
11423304 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413304 Supplier Thermo Scientific Alfa Aesar Supplier No. A10609.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2103-57-3 | C10H12O4 | 196.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2103-57-3
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003310
InChI Key UCTUXUGXIFRVGX-UHFFFAOYSA-N
Synonym trimethoxybenzaldehyde, benzaldehyde, 2,3,4-trimethoxy, 2,3,4-trimethoxy-benzaldehyde, benzaldehyde, trimethoxy, 2,3,4 trimethoxybenzylaldehyde, pubchem8261, trimethoxy benzaldehyde, trimetazidine impurity c, acmc-1b07v, ksc201q5r
PubChem CID 75006
IUPAC Name 2,3,4-trimethoxybenzaldehyde
SMILES COC1=C(C(=C(C=C1)C=O)OC)OC

Specifications

Melting Point 38°C to 42°C
Boiling Point 168°C to 170°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5547
Quantity 50 g
Beilstein 981091
Sensitivity Air Sensitive
Formula Weight 196.2
Percent Purity ≥98%
Chemical Name or Material 2,3,4-Trimethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.