missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080851
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
2103-57-3
C10H12O4
196.202
MFCD00003310
UCTUXUGXIFRVGX-UHFFFAOYSA-N
trimethoxybenzaldehyde, benzaldehyde, 2,3,4-trimethoxy, 2,3,4-trimethoxy-benzaldehyde, benzaldehyde, trimethoxy, 2,3,4 trimethoxybenzylaldehyde, pubchem8261, trimethoxy benzaldehyde, trimetazidine impurity c, acmc-1b07v, ksc201q5r
75006
2,3,4-trimethoxybenzaldehyde
COC1=C(C(=C(C=C1)C=O)OC)OC
This item is not returnable. View return policy
2103-57-3
C10H12O4
196.202
MFCD00003310
UCTUXUGXIFRVGX-UHFFFAOYSA-N
trimethoxybenzaldehyde, benzaldehyde, 2,3,4-trimethoxy, 2,3,4-trimethoxy-benzaldehyde, benzaldehyde, trimethoxy, 2,3,4 trimethoxybenzylaldehyde, pubchem8261, trimethoxy benzaldehyde, trimetazidine impurity c, acmc-1b07v, ksc201q5r
75006
2,3,4-trimethoxybenzaldehyde
COC1=C(C(=C(C=C1)C=O)OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 38°C to 42°C
Boiling Point 168°C to 170°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5547
Quantity 10 g
Beilstein 981091
Sensitivity Air Sensitive
Formula Weight 196.2
Percent Purity ≥98%
Chemical Name or Material 2,3,4-Trimethoxybenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'