missing translation for 'onlineSavingsMsg'
Learn More

[(2,3,4-Trihydroxyphenyl)methylene]hydrazide-serine Hydrochloride, TRC

Product Code. 30448978
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30448978 100 mg 100mg
30452673 250 mg 250mg
30455738 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30448978 Supplier Toronto Research Chemicals Supplier No. TRCT895300100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1353749-74-2
Molecular Formula C10H13N3O5 . HCl
Molecular Weight (g/mol) 291.69
Synonym Serine, 2-[(2,3,4-trihydroxyphenyl)methylene]hydrazide, hydrochloride (1:1), (2RS)-2-Amino-3-hydroxy-2'-[(1EZ)-(2,3,4-trihydroxybenzylidene)]propanohydrazide Hydrochloride, Benserazide Imp. C (EP) as Hydrochloride
IUPAC Name 2-amino-3-hydroxy-N-[(2,3,4-trihydroxyphenyl)methylideneamino]propanamide;hydrochloride
SMILES NC(CO)C(=O)NN=Cc1ccc(O)c(O)c1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H13N3O5.ClH/c11-6(4-14)10(18)13-12-3-5-1-2-7(15)9(17)8(5)16;/h1-3,6,14-17H,4,11H2,(H,13,18);1H
Formula Weight 291.0622
Chemical Name or Material [(2,3,4-Trihydroxyphenyl)methylene]hydrazide-serine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.