missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzoic acid, MedChemExpress

Product Code. 16459386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459386 Supplier Medchem Express Supplier No. HYW016993

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 2,3,4-Trihydroxybenzoic acid is an internal standard in separation of phenolic acids by HPLC.

2,3,4-Trihydroxybenzoic acid is an internal standard in separation of phenolic acids by HPLC.

Specifications

Chemical Name or Material 2,3,4-Trihydroxybenzoic acid
CAS 610-02-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H6O5
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (293.91 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(O)C(O)=C1O
Molecular Weight (g/mol) 170.12
Formula Weight 170.12
Percent Purity 99.22%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.