missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzoic acid, 97%

Product Code. 11494277
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494277 25 g 25g
11474277 1 g 1g
11484277 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494277 Supplier Thermo Scientific Alfa Aesar Supplier No. B20371.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610-02-6 | C7H6O5 | 170.12 g/mol

2,3,4-Trihydroxybenzoic acid is used as a reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters and used in the pharmaceuticals. It is a internal standard in separation of phenolic acids by HPLC with photodiode array detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4-Trihydroxybenzoic acid is used as a reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters and used in the pharmaceuticals. It is a internal standard in separation of phenolic acids by HPLC with photodiode array detection.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-02-6
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002447
InChI Key BRRSNXCXLSVPFC-UHFFFAOYSA-N
Synonym 4-pyrogallolcarboxylic acid, pyrogallolcarboxylic acid, 2,3,4-trihydroxybenzoate, unii-ad1id2jf5o, benzoic acid, trihydroxy, benzoic acid, 2,3,4-trihydroxy, pyrogallol-4-carboxylic acid, 2,3,4-trihydroxybenzene carboxylic acid, ad1id2jf5o, pyrogallolcarboxylate
PubChem CID 11874
IUPAC Name 2,3,4-trihydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(O)=C1O

Specifications

Melting Point ∼210°C (decomposition)
Quantity 25 g
Beilstein 2210807
Solubility Information Slightly soluble in water.
Formula Weight 170.12
Percent Purity 97%
Chemical Name or Material 2,3,4-Trihydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.