missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 11984541
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 157911-55-2
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.01
InChI Key DGSXDQVPGXFOAN-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzyl bromide, 1-bromomethyl-2,3,4-trifluorobenzene, 2,3,4-trifluorobenzylbromide, 6-bromomethyl-1,2,3-trifluorobenzene, benzene, 1-bromomethyl-2,3,4-trifluoro, alpha-bromo-2,3,4-trifluorotoluene, 2,3,4-trifluoro benzyl bromide, 2,3,4-trifluorophenyl methyl bromide, 4-bromomethyl-1,2,3-trifluorobenzene, 2, 3, 4-trifluorobenzyl bromide
PubChem CID 2777050
IUPAC Name 1-(bromomethyl)-2,3,4-trifluorobenzene
SMILES C1=CC(=C(C(=C1CBr)F)F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 225.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3, 4-Trifluorobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4-Trifluorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'