missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzyl alcohol, 97%, Thermo Scientific™

Catalog Number 11475727
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
144284-24-2
C7H5F3O
162.111
MFCD00083551
LOZFHOHUIYBNCJ-UHFFFAOYSA-N
2,3,4-trifluorophenyl methanol, 2,3,4-trifluorobenzyl alcohol, 2,3,4-trifluorobenzylalcohol, 2,3,4-trifluoro benzenemethanol, 2,3,4-trifluoro-phenyl-methanol, 2,3,4-trifluorophenyl methan-1-ol, pubchem8282, acmc-209vax, benzenemethanol,2,3,4-trifluoro, 2,3,4-tris fluoranyl phenyl methanol
2777027
(2,3,4-trifluorophenyl)methanol
C1=CC(=C(C(=C1CO)F)F)F
This item is not returnable. View return policy
144284-24-2
C7H5F3O
162.111
MFCD00083551
LOZFHOHUIYBNCJ-UHFFFAOYSA-N
2,3,4-trifluorophenyl methanol, 2,3,4-trifluorobenzyl alcohol, 2,3,4-trifluorobenzylalcohol, 2,3,4-trifluoro benzenemethanol, 2,3,4-trifluoro-phenyl-methanol, 2,3,4-trifluorophenyl methan-1-ol, pubchem8282, acmc-209vax, benzenemethanol,2,3,4-trifluoro, 2,3,4-tris fluoranyl phenyl methanol
2777027
(2,3,4-trifluorophenyl)methanol
C1=CC(=C(C(=C1CO)F)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.472
Quantity 1 g
Formula Weight 162.11
Percent Purity 97%
Chemical Name or Material 2,3,4-Trifluorobenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'