missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-2696675
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10243522 1 g 1g
10732513 10 g 10g
11356965 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10243522 Supplier Thermo Scientific Maybridge Supplier No. TL00605DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-08-7
Molecular Formula C6H2ClF3O2S
Molecular Weight (g/mol) 230.59
MDL Number MFCD00102573
InChI Key XFTDZYFHXRZLEF-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzene-1-sulfonyl chloride, 2,3,4-trifluorobenzenesulphonyl chloride, 2,3,4-trifluorophenylsulfonyl chloride, 2,3,4-trifluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,3,4-trifluoro, benzenesulfonyl chloride, 2,3,4-trifluoro-9ci, acmc-20aoih, pubchem5177, 2,3,4-trifluorobenz, buttpark 44\07-12
PubChem CID 2776977
SMILES FC1=CC=C(C(F)=C1F)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Density 1.6400g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula F3C6H2SO2Cl
Quantity 1 g
IUPAC Name 2,3,4-trifluorobenzenesulfonyl chloride
Formula Weight 230.59
Percent Purity 97%
Chemical Name or Material 2, 3, 4-Trifluorobenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.