missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzaldehyde, 98%, Thermo Scientific™

Product Code. 10201283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10201283 5 g Glass bottle 5g
10796233 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10201283 Supplier Thermo Scientific Acros Supplier No. 312710050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 161793-17-5
Molecular Formula C7H3F3O
Molecular Weight (g/mol) 160.09
MDL Number MFCD00061230
InChI Key UQEDGFZRPSAHLC-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenz-aldehyde, benzaldehyde, 2,3,4-trifluoro, 2,3,4-trifluoro benzaldehyde, pubchem4255, intermediates-zcf02215, acmc-209dn7, 2,3,4-trilfuorobenzaldehyde, ksc182o7d, 2,3,4-trifluoro-benzaldehyde, timtec-bb sbb003969
PubChem CID 519226
IUPAC Name 2,3,4-trifluorobenzaldehyde
SMILES C1=CC(=C(C(=C1C=O)F)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.4040g/mL
Boiling Point 77°C to 79°C (20.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F3C6H2CHO
Refractive Index 1.4800 to 1.482
Quantity 5 g
Specific Gravity 1.404
Formula Weight 160.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3, 4-Trifluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.