missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluoroaniline, 98%

Product Code. 11463963
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463963 25 g 25g
11453963 5 g 5g
11473963 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463963 Supplier Thermo Scientific Alfa Aesar Supplier No. A10254.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3862-73-5 | C6H4F3N | 147.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3862-73-5
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00011737
InChI Key WRDGNXCXTDDYBZ-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzenamine, benzenamine, 2,3,4-trifluoro, 2,3,4-trifluoraniline, 2,3,4-trifluoro-phenylamine, aniline, 2,3,4-trifluoro, 2,3,4-trifluoro aniline, 2,3,4-trifluorophenylamine, pubchem2297, acmc-1clgf, 2.3.4-trifluoroaniline
PubChem CID 77468
IUPAC Name 2,3,4-trifluoroaniline
SMILES C1=CC(=C(C(=C1N)F)F)F

Specifications

Density 1.393
Boiling Point 90°C to 92°C (48 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.487
Quantity 25 g
UN Number UN3082
Beilstein 3245609
Formula Weight 147.1
Percent Purity 98%
Chemical Name or Material 2,3,4-Trifluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.