missing translation for 'onlineSavingsMsg'
Learn More

2',3',4'-Trichloroacetophenone, 95%, Thermo Scientific™

Product Code. 11444864
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444864 100 g 100g
11424864 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444864 Supplier Thermo Scientific Alfa Aesar Supplier No. A13790.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13608-87-2
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.477
MDL Number MFCD00000542
InChI Key BXJZZJYNVIDEKG-UHFFFAOYSA-N
Synonym 2',3',4'-trichloroacetophenone, 1-2,3,4-trichlorophenyl ethanone, 2,3,4-trichloroacetophenone, ethanone, 1-2,3,4-trichlorophenyl, 1-2,3,4-trichlorophenyl ethan-1-one, ccris 785, unii-8hvd33n785, acetophenone, 2',3',4'-trichloro, ethanone,1-2,3,4-trichlorophenyl, 1-acetyl-2,3,4-trichlorobenzene
PubChem CID 26132
IUPAC Name 1-(2,3,4-trichlorophenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)Cl)Cl)Cl

Specifications

Melting Point 60°C to 64°C
Quantity 100 g
Beilstein 1948674
Formula Weight 223.49
Percent Purity 95%
Chemical Name or Material 2',3',4'-Trichloroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.