missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dimethoxyphenyl)-N-methylethylamine, TRC

Product Code. 30383311
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383311 10 g
30370778 1 g
30383989 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383311 Supplier Toronto Research Chemicals Supplier No. TRCD47071010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3490-06-0
Molecular Formula C11 H17 N O2
Molecular Weight (g/mol) 195.26
Synonym Benzeneethanamine, 3,4-dimethoxy-N-methyl- (9CI, ACI), 3,4-Dimethoxy-N-methylbenzeneethanamine (ACI), Phenethylamine, 3,4-dimethoxy-N-methyl- (6CI, 7CI, 8CI), 2-(3,4-Dimethoxyphenyl)-N-methylethanamine, 2-(3,4-Dimethoxyphenyl)-N-methylethylamine, 3,4-Dimethoxy-N-methylphenethylamine, 3,4-Dimethoxy-N-methylphenylethylamine, 3,4-Dimethoxy-β-phenethylmethylamine, [2-(3,4-Dimethoxyphenyl)ethyl]methylamine, Methyl[2-(3,4-dimethoxyphenyl)ethyl]amine, N-(3,4-Dimethoxyphenethyl)-N-methylamine, N-[2-(3,4-Dimethoxyphenyl)ethyl]-N-methylamine, N-Methyl-2-(3,4-dimethoxyphenyl)ethylamine, N-Methyl-3,4-dimethoxy-β-phenethylamine, N-Methyl-3,4-dimethoxy-β-phenylethylamine, N-Methyl-3,4-dimethoxybenzeneethanamine, N-Methyl-3,4-dimethoxyphenethylamine, N-Methylhomoveratrylamine, NSC 187772
IUPAC Name 2-(3,4-dimethoxyphenyl)-N-methylethanamine
SMILES CNCCc1ccc(OC)c(OC)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H17NO2/c1-12-7-6-9-4-5-10(13-2)11(8-9)14-3/h4-5,8,12H,6-7H2,1-3H3
Formula Weight 195.1259
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)-N-methylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.