missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11352637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352637 1 g 1g
11362637 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352637 Supplier Thermo Scientific Alfa Aesar Supplier No. L11817.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21714-25-0
Molecular Formula C11H13NO2S
Molecular Weight (g/mol) 223.29
MDL Number MFCD00041366
InChI Key GHYFLDWHIVKQLS-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
PubChem CID 140856
IUPAC Name 4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
SMILES COC1=CC=C(CCN=C=S)C=C1OC

Specifications

Density 1.17
Boiling Point 150°C to 151°C (0.3 mmHg)
Odor Irritating
Refractive Index 1.588
Quantity 1 g
UN Number UN2922
Beilstein 2809316
Sensitivity Moisture sensitive
Formula Weight 223.3
Percent Purity 98%
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.