missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-benzyl-Alpha-D-glucopyranosyl Bromide 90%Stabilized with Calcium Carbonate, TRC

Product Code. 30416427
Change view
Click to view available options
Quantity:
500 mg
2.5 g
5 g
Unit Size:
2.5g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416427 5 g 5g
30415779 500 mg 500mg
30419209 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416427 Supplier Toronto Research Chemicals Supplier No. TRCT2892055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4196-35-4
Molecular Formula C34H35BrO5
Molecular Weight (g/mol) 603.54
Synonym 2,3,4,6-Tetrakis-O-(phenylmethyl)-alpha-D-glucopyranosyl Bromide,, 2,3,4,6-Tetra-O-benzyl-glucopyranosyl Bromide, alpha-D-2,3,4,6-Tetra-O-benzyl-glucopyranosyl Bromide, 2,3,4,6-Tetra-O-benzyl-1-bromo-1-deoxy-alpha-D-glucopyranose, 2,3,4,6-Tetra-O-benzyl-alpha-D-bromoglucopyranoside, 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl Bromide, Tetra-O-benzyl-alpha-D-glucopyranosyl Bromide
IUPAC Name (2R,3R,4S,5R,6R)-2-bromo-3,4,5-tris(phenylmethoxy)-6-(3-phenylpropyl)oxane
SMILES Br[C@H]1O[C@H](CCCc2ccccc2)[C@@H](OCc3ccccc3)[C@H](OCc4ccccc4)[C@H]1OCc5ccccc5

Specifications

Quantity 5 g
InChI Formula InChI=1S/C35H37BrO4/c36-35-34(39-26-30-20-11-4-12-21-30)33(38-25-29-18-9-3-10-19-29)32(37-24-28-16-7-2-8-17-28)31(40-35)23-13-22-27-14-5-1-6-15-27/h1-12,14-21,31-35H,13,22-26H2/t31-,32-,33+,34-,35+/m1/s1
Formula Weight 600.1875
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.