missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3,4,6-Tetra-O-acetyl-D-glucopyranose, 98%

Product Code. 15497259
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497259 5 g 5g
15487259 1 g 1g
15407269 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497259 Supplier Thermo Scientific Chemicals Supplier No. J66052.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10343-06-3 | C14H20O10 | 348.304 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform or ethyl acetate

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10343-06-3
Molecular Formula C14H20O10
Molecular Weight (g/mol) 348.304
MDL Number MFCD00061634
InChI Key IEOLRPPTIGNUNP-RQICVUQASA-N
Synonym 2,3,4,6-tetra-o-acetyl-d-glucopyranose, 2,3,4,6-tetra-o-acetylglucopyranose, 2r,3r,4s,5r-3,4,5-tris acetyloxy-6-hydroxyoxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-beta-d-glucopyranose, pubchem10600, d-glucopyranose 2,3,4,6-tetraacetate, 2,3,4,6-tetra-o-acetyl-glucopyranose, 2,3,4,6-tetraacetate d-glucopyranose, d-glucopyranose, 2,3,4,6-tetraacetate, 2r,3r,4s,5r-2-acetoxymethyl-6-hydroxytetrahydro-2h-pyran-3,4,5-triyl triacetate
PubChem CID 11824227
IUPAC Name [(2R,3R,4S,5R)-3,4,5-triacetyloxy-6-hydroxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)O)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 137°C to 138°C
Color White
Quantity 5 g
Solubility Information Soluble in chloroform or ethyl acetate
Formula Weight 348.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-D-glucopyranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.