missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%

Product Code. 10666851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10666851 10 g Glass bottle 10g
10269153 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10666851 Supplier Thermo Scientific Acros Supplier No. 270930100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 572-09-8 | C14H19BrO9 | 411.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 572-09-8
Molecular Formula C14H19BrO9
Molecular Weight (g/mol) 411.20
MDL Number MFCD00063254
InChI Key CYAYKKUWALRRPA-HAGGZJDPNA-N
Synonym 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromoglucose, acetobromo-alpha-d-glucose, a-acetobromglucose, acetobromo glucose, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, stab. with calcium carbonate
PubChem CID 101776
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 83.0°C to 88.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Argentometry)
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 59
Specific Rotation Condition +194.5° (20°C c=2,chloroform)
Specific Rotation + 194.50
Solubility Information Solubility in water: decomposes. Other solubilities: 1g/20mL abs. ethanol - more soluble in methanol, freely soluble in ether, chloroform, acetone,, ethyl acetate and benzene, slightly soluble in petroleum ether
Stabilizer 2% CaCO3
Formula Weight 411.2
Percent Purity 98.5%
Physical Form Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4,6-Tetra-O-acetyl-a-D-glucopyranosyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.