missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluorobenzoic acid, 98+%, Thermo Scientific™

Catalog Number 11413198
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1201-31-6
C7H2F4O2
194.09
MFCD00009613
SFKRXQKJTIYUAG-UHFFFAOYSA-N
2,3,4,5-tetrafluoro benzoic acid, benzoic acid, tetrafluoro, pubchem1350, tetrafluorobenzoic acid, acmc-1bre4, ksc174o7d, sfkrxqkjtiyuag-uhfffaoysa, 2,3,4,5-tetrafluorbenzoic acid, 2,3,4,5-tetrafluoro-benzoic acid, 2,3,4,5-?tetrafluorobenzoic acid
297549
2,3,4,5-tetrafluorobenzoic acid
OC(=O)C1=CC(F)=C(F)C(F)=C1F
This item is not returnable. View return policy
1201-31-6
C7H2F4O2
194.09
MFCD00009613
SFKRXQKJTIYUAG-UHFFFAOYSA-N
2,3,4,5-tetrafluoro benzoic acid, benzoic acid, tetrafluoro, pubchem1350, tetrafluorobenzoic acid, acmc-1bre4, ksc174o7d, sfkrxqkjtiyuag-uhfffaoysa, 2,3,4,5-tetrafluorbenzoic acid, 2,3,4,5-tetrafluoro-benzoic acid, 2,3,4,5-?tetrafluorobenzoic acid
297549
2,3,4,5-tetrafluorobenzoic acid
OC(=O)C1=CC(F)=C(F)C(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 85°C to 87°C
Quantity 1 g
Beilstein 2052670
Formula Weight 194.09
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'