missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-Di-O-isopropylidene-D-arabitol, 98%, Thermo Scientific Chemicals

Product Code. 15488029
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488029 1 g 1g
15498029 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488029 Supplier Thermo Scientific Alfa Aesar Supplier No. J66608.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19139-74-3
Molecular Formula C11H20O5
Molecular Weight (g/mol) 232.28
MDL Number MFCD00063387
InChI Key FBIWGPWXTDOFQR-UHFFFAOYNA-N
Synonym 2-o,3-o:4-o,5-o-diisopropylidene-d-arabinitol
PubChem CID 11042611
IUPAC Name [(4R,5R)-5-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(C)OCC(O1)C1OC(C)(C)OC1CO

Specifications

Color Colorless
Quantity 1 g
Formula Weight 232.27
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 2,3:4,5-Di-O-isopropylidene-D-arabitol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.